  • Search Results
  • A protein kinase, calcineurin B-like protein-interacting protein Kinase9, interacts with calcium sensor calcineurin B-like Protein3 and regulates potassium homeostasis under low-potassium stress in Arabidopsis.
A protein kinase, calcineurin B-like protein-interacting protein Kinase9, interacts with calcium sensor calcineurin B-like Protein3 and regulates potassium homeostasis under low-potassium stress in Arabidopsis.
Plant physiology (2012-10-31)
Li-Li Liu, Hui-Min Ren, Li-Qing Chen, Yi Wang, Wei-Hua Wu
ABSTRACT
Potassium (K⁺) is an essential macronutrient for plant growth and development. Previous studies have demonstrated that Calcineurin B-Like Protein1 (CBL1) or CBL9 and CBL-Interacting Protein Kinase23 (CIPK23) regulate K⁺ uptake in Arabidopsis (Arabidopsis thaliana) roots by modulating K⁺ channel Arabidopsis K⁺ Transporter1. In this study, we show that the protein kinase CIPK9 interacts with the calcium sensor CBL3 and plays crucial roles in K⁺ homeostasis under low-K⁺ stress in Arabidopsis. Arabidopsis wild-type plants showed leaf chlorotic symptoms when grown for 10 d on low-K⁺ (100 μM) medium. Here, we show that plants lacking CIPK9 displayed a tolerant phenotype to low-K⁺ stress, which still maintained green leaves when the wild-type plants showed typical K⁺-deficient symptoms. Overexpressing lines of CIPK9 resulted in a low-K⁺-sensitive phenotype compared with wild-type plants. Furthermore, CBL2 and CBL3 were identified as upstream regulators of CIPK9. Both CBL2- and CBL3-overexpressing lines displayed similar low-K⁺-sensitive phenotypes and K⁺ contents to CIPK9-overexpressing lines. However, only cbl3 mutant plants, but not cbl2 mutant plants, showed the low-K⁺-tolerant phenotype similar to cipk9 mutants. Taken together, these results demonstrate that CIPK9 and CBL3 work together and function in K⁺ homeostasis under low-K⁺ stress in Arabidopsis.
Sigma-Aldrich
Protein Kinase A from bovine heart, ≥0.4 units/μg protein, lyophilized powder
Sigma-Aldrich
Protein Kinase CβI isozyme human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein Kinase Cζ isozyme human, ≥75% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution
Sigma-Aldrich
Protein kinase CβII isozyme human, >85% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein Kinase Cδ isozyme human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein kinase Cγ isozyme human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution