  • Search Results
  • [Utility of chromosome banding with ALU I enzyme for identifying methylated areas in breast cancer].
[Utility of chromosome banding with ALU I enzyme for identifying methylated areas in breast cancer].
Investigacion clinica (2013-03-22)
Alicia Rojas-Atencio, Leonard Yamarte, Karelis Urdaneta, Marisol Soto-Alvarez, Francisco Alvarez Nava, Jenny Cañizalez, Maribel Quintero, Raquel Atencio, Richard González
ABSTRACT
Cancer is a group of disorders characterized by uncontrolled cell growth which is produced by two successive events: increased cell proliferation (tumor or neoplasia) and the invasive capacity of these cells (metastasis). DNA methylation is an epigenetic process which has been involved as an important pathogenic factor of cancer. DNA methylation participates in the regulation of gene expression, directly, by preventing the union of transcription factors, and indirectly, by promoting the "closed" structure of the chromatine. The objectives of this study were to identify hypermethyled chromosomal regions through the use of restriction Alu I endonuclease, and to relate cytogenetically these regions with tumor suppressive gene loci. Sixty peripheral blood samples of females with breast cancer were analyzed. Cell cultures were performed and cytogenetic spreads, previously digested with Alu I enzyme, were stained with Giemsa. Chromosomal centromeric and not centromeric regions were stained in 37% of cases. About 96% of stained hypermethyled chromosomal regions (1q, 2q, 6q) were linked with methylated genes associated with breast cancer. In addition, centromeric regions in chromosomes 3, 4, 8, 13, 14, 15 and 17, usually unstained, were found positive to digestion with Alu I enzime and Giemsa staining. We suggest the importance of this technique for the global visualization of the genome which can find methylated genes related to breast cancer, and thus lead to a specific therapy, and therefore a better therapeutic response.
Sigma-Aldrich
Nde I from Neisseria denitrificans, Restriction Enzyme