  • Search Results
  • Construction, expression and characterisation of recombinant molecules of the urease gene operon from a urease-positive thermophilic Campylobacter (UPTC) isolate.
Construction, expression and characterisation of recombinant molecules of the urease gene operon from a urease-positive thermophilic Campylobacter (UPTC) isolate.
British journal of biomedical science (2013-04-27)
S Nakanishi, T Nakajima, A Tazumi, K Matsubara, J E Moore, B C Millar, M Matsuda
ABSTRACT
A recombinant molecule of the full-length urease gene operon was constructed in vitro from the Japanese urease-positive thermophilic Campylobacter (UPTC) CF89-12 isolate and expressed in Escherichia coli cells. Several large deletion recombinant variants of urease subunit genes were also constructed and expressed in E. coli cells. A positive urease reaction with the log-phase cultured E. coli JM109 cells in the NiCl2-containing medium transformed with pGEM-T vector carrying the recombinant molecule of the full-length operon was detected with isopropyl-beta-D-thiogalactoside. Among the several deletion recombinant variants, each ureA-, ureB-, ureE-, ureF-, ureG- and ureH-large deficient, only ureE-large deletion variant (63% deficient) showed a positive urease reaction (approximately 15-fold). In addition, a ureE-complete deletion recombinant variant (100% deficient) constructed also showed a positive reaction of urease (approximately 18-fold). Recombinant urease subunits A and B were immunologically identified by Western blot analysis with anti-urease alpha (A) and beta (B) raised against Helicobacter pylori.
Sigma-Aldrich
Nickel(II) chloride, 98%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Nickel(II) chloride hexahydrate, ReagentPlus®
Sigma-Aldrich
Nickel(II) chloride hexahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Nickel(II) chloride hexahydrate, puriss. p.a., ≥98%
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg
Sigma-Aldrich
Nickel(II) chloride hydrate, 99.95% trace metals basis