  • Search Results
  • [Mutation analysis of the HBV reverse transcriptase in nucleos(t)ide-treated patients with chronic HBV infection].
[Mutation analysis of the HBV reverse transcriptase in nucleos(t)ide-treated patients with chronic HBV infection].
Zhonghua shi yan he lin chuang bing du xue za zhi = Zhonghua shiyan he linchuang bingduxue zazhi = Chinese journal of experimental and clinical virology (2013-05-01)
Xiu-Juan Jiao, Xun Peng, Xiu-Min Jiao, Jin-Sheng Wang, Xie-Wen Sun, Pei-Li Zhao, Shou-Yun Wang, Jia-Qun Liu, Tong Li, Jing-Xian Yang
ABSTRACT
To characterize genotypic resistance within HBV RT region in chronic hepatitis B (CHB) patients with nucleos(t)ide analogue (NA) treatment. Serum samples of 229 CHB patients with NA treatment were obtained. Full-length HBV RT sequences were amplified, sequenced and analyzed, on the following NA resistant (NAr) mutations belonging to different NAr pathways. Among 229 HBV isolates, 14.41% (33/229) and 85.59% (196/229) were genotype B and C, respectively; and the patients with HBV genotype C may be more susceptible to develope resistant mutations than patients with HBV genotype B(chi2 = 2.95, P < 0.05). NAr mutations were detected in 63 CHB patients. Mutations were not found at rtI169, rtT184, rtA194 or rtS202. RtM204 mutations were detected at the highest frequency among 63 mutants (40/63, 63.49%) and found to display 11 combination mutation patterns, in which rtM204I were associated with rtL80I/V and rtL180M, and rtM204V were associated with rtL1l80M, respectively. Conclusions There are complicated mutation patterns in the HBV RT region for chronic hepatitis B (CHB) patients with nucleos(t)ide analogue (NA) treatment. RtM204V/I mutation was the highest.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.