  • Search Results
  • Homeodomain-interacting protein kinase regulates Yorkie activity to promote tissue growth.
Homeodomain-interacting protein kinase regulates Yorkie activity to promote tissue growth.
Current biology : CB (2012-07-31)
Joanna Chen, Esther M Verheyen
ABSTRACT
The Hippo (Hpo) tumor suppressor pathway regulates tissue size by inhibiting cell proliferation and promoting apoptosis. The core components of the pathway, Hpo, Salvador, Warts (Wts), and Mats, form a kinase cascade to inhibit the activity of Yorkie (Yki), the transcriptional effector of the pathway. Homeodomain-interacting protein kinases (Hipks) are a family of conserved serine/threonine kinases that function as regulators of various transcription factors to regulate developmental processes including proliferation, differentiation, and apoptosis. Hipk can induce tissue overgrowth in Drosophila. We demonstrate that Hipk is required to promote Yki activity. Hipk affects neither Yki stability nor its subcellular localization. Moreover, hipk knockdown suppresses the overgrowth and target gene expression caused by hyperactive Yki. Hipk phosphorylates Yki and in vivo analyses show that Hipk's regulation of Yki is kinase-dependent. To our knowledge, this is the first kinase identified to positively regulate Yki.
Sigma-Aldrich
Protein Kinase A from bovine heart, ≥0.4 units/μg protein, lyophilized powder
Sigma-Aldrich
Protein Kinase CβI isozyme human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein Kinase Cζ isozyme human, ≥75% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution
Sigma-Aldrich
Protein kinase CβII isozyme human, >85% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein Kinase Cδ isozyme human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Sigma-Aldrich
Protein kinase Cγ isozyme human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution