  • Search Results
  • Antibodies to presenilin proteins detect neurofibrillary tangles in Alzheimer's disease.
Antibodies to presenilin proteins detect neurofibrillary tangles in Alzheimer's disease.
The American journal of pathology (1996-12-01)
G M Murphy, L S Forno, W G Ellis, D Nochlin, E Levy-Lahad, P Poorkaj, T D Bird, Z Jiang, B Cordell
ABSTRACT
Mutations in the presenilin (PS)-1 and PS-2 genes have been shown to be linked with the development of Alzheimer's disease (AD). We examined Alzheimer's brain tissue by immunohistochemistry using a set of antibodies raised to sequences shared between PS-1 and PS-2 proteins. These antibodies reacted exclusively with a subset of neurofibrillary tangles and not with neuropil threads or dystrophic neurites. Detection of the presenilin epitope in neurofibrillary tangles was observed in sporadic Alzheimer's disease brain samples and in samples from individuals carrying PS-1 and PS-2 mutations with no qualitative difference. These data indicate that both wild-type and mutant PS proteins are involved in a common pathogenic pathway in AD.
Sigma-Aldrich
Anti-Presenilin-1 (14-33) antibody produced in goat, whole antiserum
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.