  • Search Results
  • CT60 single nucleotide polymorphism of the CTLA-4 gene is associated with susceptibility to Graves' disease in the Taiwanese population.
CT60 single nucleotide polymorphism of the CTLA-4 gene is associated with susceptibility to Graves' disease in the Taiwanese population.
Annals of clinical and laboratory science (2005-08-06)
Yu-Ching Weng, Ming-Jiuan Wu, Wei-Sen Lin
ABSTRACT
Graves' disease (GD) is an autoimmune disease but the underlying etiology has not been completely elucidated. Genetic susceptibility has been believed to play a major role. Recent studies showed that the CT60 single nucleotide polymorphism (SNP), which is in the 3'-noncoding region of the CTLA-4 gene, is strongly associated with some immune-mediated diseases. The aim of this study was to test for association between GD susceptibility and polymorphisms of CTLA-4 (ie, the CT60 SNP and the exon 1 +49 SNP) in the Taiwanese population. Our results demonstrate significant differences in the frequencies of the genotypes and alleles between 107 GD patients and 101 control subjects in the CT60 and exon 1 +49 SNPs (p <0.05). Significant differences in phenotypes were only found for CT60 SNP (78.4% vs 67.8% between patients and controls; chi2 = 3.93, p = 0.047). Furthermore, we found that the G/G genotype of both CT60 and exon 1 +49 was associated with increased risk for GD (p = 0.022, OR = 1.97). Significant linkage disequilibrium was found between the CT60 SNP and the exon 1 +49 SNP in both GD patients and control subjects (D' = 1.00). Because of tight linkage disequilibrium, a combination of these SNPs enhanced the role of the CTLA-4 gene in GD. The frequency of the disease-susceptible G allele of CT60 was comparable to that in Japanese and higher than in Caucasians. In conclusion, we provide evidence that CT60 SNP is associated with susceptibility to GD in the Taiwanese population.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.