Screening of lipase inhibitors from marine algae.
Lipids (1999-06-24)
N Bitou, M Ninomiya, T Tsujita, H Okuda
ABSTRACT
The possible presence of an inhibitor of pancreatic lipase (triacylglycerol acylhydrolase, EC 3.1.1.3) was screened in 54 marine algae. An active inhibitor, caulerpenyne, was purified from an extract of Caulerpa taxifolia, using ethyl acetate extraction, followed by successive chromatographies on ODS and silica gel columns. The purified inhibitor was identified by thin-layer chromatography, infrared and nuclear magnetic resonance spectroscopy. Caulerpenyne competitively inhibited lipase activities using emulsified triolein and dispersed 4-methylumbelliferyl oleate (4-MU oleate) as substrates. The concentrations producing 50% inhibition against triolein and 4-MU oleate hydrolysis were 2 mM and 13 microM, respectively. In vivo, oral administration of corn oil with or without caulerpenyne to rats demonstrated a reduced and delayed peak plasma triacylglycerol concentration with caulerpenyne.
Sigma-Aldrich
4-Methylumbelliferyl oleate, suitable for fluorescence, ≥95% (HPCE)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.