  • Search Results
  • Increased levels of oxidatively damaged DNA induced by chromium(III) and H2O2: protection by melatonin and related molecules.
Increased levels of oxidatively damaged DNA induced by chromium(III) and H2O2: protection by melatonin and related molecules.
Journal of pineal research (2000-08-19)
W Qi, R J Reiter, D X Tan, L C Manchester, A W Siu, J J Garcia
ABSTRACT
Chromium (Cr) compounds are known occupational and environmental carcinogens. This trace element is found in the workplace primarily in the valence forms Cr(III) and Cr(VI). Cr(III), which was thought originally to be relatively nontoxic, was recently found to be more reactive toward purified DNA than was chromium(VI). Herein, we examined the ability of Cr(III) to induce oxidative DNA damage by measuring the formation of 8-hydroxydeoxyguanosine (8-OH-dG) in purified calf thymus DNA incubated with CrCl3 plus H2O2. In this system we observed that the Cr(III)-induced formation of 8-OH-dG in isolated DNA was both dose- and time-dependent. When melatonin and related molecules, including 6-methoxy-1,2,3,4-tetrahydro-beta-carboline (pinoline), N-acetylserotonin, 6-hydroxymelatonin and indole-3-propionic acid, were co-incubated with CrCl3 plus H2O2, the accumulations of 8-OH-dG in DNA samples were markedly inhibited in a concentration-dependent manner. The concentrations of each indole required to reduce DNA damage by 50%, i.e. the IC50 values, were 0.48, 0.51, 0.88, 1.00 and 3.08 microM for pinoline, melatonin, N-acetylserotonin, 6-hydroxymelatonin and indole-3-propionic acid, respectively. These results suggest that one of the mechanisms by which Cr(III) may induce cancer is via Fenton-type reactions which generate the hydroxyl radical (*OH). The findings also indicate that the protective effects of melatonin and related molecules against Cr(III)-induced carcinogenesis relate to their direct *OH scavenging ability which thereby reduces the formation of the damaged DNA product, 8-OH-dG.
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Sigma-Aldrich
6-Methoxy-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole, 97%