  • Search Results
  • Human tear lipocalin acts as an oxidative-stress-induced scavenger of potentially harmful lipid peroxidation products in a cell culture system.
Human tear lipocalin acts as an oxidative-stress-induced scavenger of potentially harmful lipid peroxidation products in a cell culture system.
The Biochemical journal (2001-05-05)
M Lechner, P Wojnar, B Redl
ABSTRACT
Human tear lipocalin [lipocalin 1 (lcn-1); von Ebner's gland protein] is a member of the lipocalin superfamily that is known to bind an unusual variety of lipophilic ligands. Because of its properties and its tissue-specific expression it has been suggested that lcn-1 might act as a physiological protection factor of epithelia. Overexpression of lcn-1 under certain disease conditions supported such a function. However, experimental investigations into its exact biological role and its mode of expression were impeded because lcn-1 was previously found to be produced only in serous glands. To overcome this problem we therefore sought a cell line that produced lcn-1 endogenously. Using reverse-transcriptase-mediated PCR analysis we found expression of lcn-1 in the human teratocarcinoma-derived NT2 precursor cells. Under normal conditions the production of lcn-1 is low. However, treatment of the cells with H(2)O(2) or FeSO(4), which typically induce lipid peroxidation, significantly enhanced the expression of lcn-1. Binding studies revealed that arachidonic acid and several lipid peroxidation products including 7beta-hydroxycholesterol, 8-isoprostane and 13-hydroxy-9,11-octadecadienoic acid specifically bind to lcn-1. To investigate the physiological consequence of this observation we purified holo-(lcn-1) from culture medium and extracted the bound ligands. The presence of F(2)-isoprostanes in the extracts obtained from the fractions containing lcn-1 indicates that these typical lipid peroxidation products are indeed ligands of the protein in vivo. These results support the idea that lcn-1 acts as a physiological scavenger of potentially harmful lipophilic molecules; lcn-1 might therefore be a novel member of the cellular defence against the deleterious effects of oxidative stress.
Sigma-Aldrich
11-[5-(Dimethylamino)-1-naphthalenesulfonylamino]undecanoic acid, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.