  • Search Results
  • Relevance of Frank's solvent classification as typically aqueous and typically non-aqueous to activities of firefly luciferase, alcohol dehydrogenase, and alpha-chymotrypsin in aqueous binaries.
Relevance of Frank's solvent classification as typically aqueous and typically non-aqueous to activities of firefly luciferase, alcohol dehydrogenase, and alpha-chymotrypsin in aqueous binaries.
Archives of biochemistry and biophysics (2004-12-08)
Nitin Wasantrao Fadnavis, Ramanujam Seshadri, Gurrala Sheelu, Kallakunta Vasantha Madhuri
ABSTRACT
Effects of cosolvent concentration on activity of fire fly luciferase, alpha-chymotrypsin, and alcohol dehydrogenase from baker's yeast (Saccharomyces cerevisiae) have been studied for several solvents with varying hydrophobicities (logP from +1.0 to -1.65) and polarities (dielectric constant from 7.4 to 109). The inhibitory effect of the cosolvent is examined in light of Frank's classification of solvents into 'typically aqueous (TA)' and 'typically non-aqueous (TNA).' The solvent concentration at which the enzyme activity decreases to half, the C(50) values, for TA solvents such as 1-cyclohexyl-2-pyrrolidinone, 2-butoxyethanol, 1-methyl-2-pyrrolidinone, tetrahydrofuran, t-butanol, and ethanol correlate quite well with their critical hydrophobic interaction concentration, rather than logP, while those for TNA solvents such as acetonitrile, dimethyl formamide, formamide, and dimethyl sulfoxide correlate well with logP. The interactions of TA solvents with proteins appear to be governed mainly by hydrophobic interactions while both hydrophobic and hydrophilic interactions play important role in case of TNA solvents.
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Supelco
1-Methyl-2-pyrrolidinone, analytical standard