  • Search Results
  • Fumigant distribution in forest nursery soils under water seal and plastic film after application of dazomet, metam-sodium and chloropicrin.
Fumigant distribution in forest nursery soils under water seal and plastic film after application of dazomet, metam-sodium and chloropicrin.
Pest management science (2006-02-14)
Dong Wang, Stephen W Fraedrich, Jennifer Juzwik, Kurt Spokas, Yi Zhang, William C Koskinen
ABSTRACT
Adequate concentration, exposure time and distribution uniformity of activated fumigant gases are prerequisites for successful soil fumigation. Field experiments were conducted to evaluate gas phase distributions of methyl isothiocyanate (MITC) and chloropicrin (CP) in two forest-tree nurseries. Concentrations of MITC and CP in soil air were measured from replicated microplots that received dazomet, metam-sodium and CP. Half of the plots were covered with high-density polyethylene tarp immediately after fumigation; the other half were not covered but received daily sprinkler irrigation for 1 week to create and maintain a water seal. The magnitude of MITC concentrations was similar between nurseries for metam-sodium in both tarp and water seal treatments and for dazomet in the tarp treatment. Consistently greater MITC and CP concentrations were found in the upper 30 cm of soil in the tarped plots compared with the water-sealed plots. Despite potential environmental and economic benefits with the water seal method, tarp covers were more reliable for achieving and maintaining higher MITC and CP concentrations and less prone to variations due to irrigation/rain, soil bulk density and other environmental conditions.
Sigma-Aldrich
Methyl isothiocyanate, 97%
Supelco
Metam-sodium hydrate, PESTANAL®, analytical standard
Supelco
Dazomet, PESTANAL®, analytical standard
Supelco
Methyl isothiocyanate, PESTANAL®, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.