MilliporeSigma
  • Liquid chromatographic determination of N-methyl carbamate pesticide residues at low parts-per-billion levels in eggs.
Liquid chromatographic determination of N-methyl carbamate pesticide residues at low parts-per-billion levels in eggs.
Journal of AOAC International (2006-03-04)
Frank J Schenck, Lynda V Podhorniak, James Hobbs, John Casanova, Dan Donoghue
ABSTRACT
A reversed-phase liquid chromatographic (LC) method is presented for the analysis of N-methyl carbamate pesticide residues and piperonyl butoxide in eggs at levels as low as 2 microg/kg (ppb). The study was undertaken to provide data for dietary exposure estimates used in risk analysis. The method uses an acetonitrile extraction followed by liquid-liquid partitioning and normal-phase aminopropyl solid-phase extraction column cleanup. Determination of residues is by reversed-phase LC with an inline postcolumn reaction followed by fluorescence detection. The average recoveries of 21 fortified (most at 2.0 and 20.0 ppb) N-methyl carbamate pesticide residues and the carbamate metabolite 1-naphthol from eggs ranged from 70 to 107%. Recoveries of the pesticide synergist piperonyl butoxide ranged from 63 to 106%. Single-comb White Leghorn hens were treated with the carbamate carbaryl, and the eggs subsequently produced were analyzed for carbaryl and 1-naphthol residues.
Sigma-Aldrich
Methyl carbamate, 98%