  • Search Results
  • Removal of the 2.2.2 cryptand (Kryptofix 2.2.2) from 18FDG by cation exchange.
Removal of the 2.2.2 cryptand (Kryptofix 2.2.2) from 18FDG by cation exchange.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes (1991-01-01)
D L Alexoff, J S Fowler, S J Gatley
ABSTRACT
The 2.2.2 cryptand [4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo-(8.8.8)-hexacosane] was trapped efficiently on Dowex AG50W-X8 (100-200 mesh) cation exchange resin. The concentration of the 2.2.2 cryptand in water with 0.1-2% methanol, or in 1 N HCl, was decreased by a factor of > 4000 using 1 mL (1.7 mequiv.) of AG50W-X8 and a flow rate of approx. 2 mL/min. K+, Cs+, Ag+ and Ba2+ forms were about equal in their ability to remove the 2.2.2 cryptand. A disposable cartridge containing 1 mL of hydrogen form resin was inserted into our automated 18FDG system so that the hydrolysate (in 2 cm3 of 1 N HCl) would pass through the cartridge before final purification. No cryptand was detected in the final product as determined by TLC with idoplantinate visualization. The detection limit was 2.5 micrograms/mL. Less than 3% of the total starting radioactivity was retained by the cation column. Quality assurance tests including apyrogenicity, sterility, radiochemical purity, carbohydrate composition and pH were not compromised by the incorporation of the cryptand removal cartridge.
Sigma-Aldrich
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, 98%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.