  • Search Results
  • Distribution and efficacy of drip-applied metam-sodium against the survival of Rhizoctonia solani and yellow nutsedge in plastic-mulched sandy soil beds.
Distribution and efficacy of drip-applied metam-sodium against the survival of Rhizoctonia solani and yellow nutsedge in plastic-mulched sandy soil beds.
Pest management science (2007-04-03)
Byron L Candole, Alexander S Csinos, Dong Wang
ABSTRACT
The effects of metam-sodium application rate on soil residence time, spatial and temporal distributions of methyl isothiocyanate and pest control efficacy were studied in a Georgia sandy soil. Metam-sodium 420 g L(-1) SL was drip applied at rates of 147 and 295 L ha(-1) in plastic-mulched raised beds. Methyl isothiocyanate concentrations in soil air space were monitored from four preselected sites: 10 and 20 cm below the emitter, and 20 and 30 cm laterally away from the emitter at 3, 12, 24, 48, 72, 120 and 240 h after chemigation. A higher rate of metam-sodium application resulted in higher methyl isothiocyanate concentrations in the soil. Highest methyl isothiocyanate concentrations were found at 20 cm below the emitter, and lowest at 30 cm laterally away from the emitter. Methyl isothiocyanate concentrations decreased with time and distance from the emitter. Lower methyl isothiocyanate concentration x time product values at 20 and 30 cm away from the emitter resulted in lower mortalities of Rhizoctonia solani Kühn and yellow nutsedge (Cyperus esculentus L.). The results demonstrated that methyl isothiocyanate can be delivered at lethal doses with drip-applied water downward within the beds. Lateral diffusion of methyl isothiocyanate from the point of application did not reach biologically active concentrations to affect the survival of R. solani or yellow nutsedge. Further studies on the lateral distribution of methyl isothiocyanate in sandy soils are needed to circumvent this limitation.
Sigma-Aldrich
Methyl isothiocyanate, 97%
Supelco
Metam-sodium hydrate, PESTANAL®, analytical standard
Supelco
Methyl isothiocyanate, PESTANAL®, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.