  • Search Results
  • Addition to electron deficient olefins of alpha-oxy carbon- centered radicals, generated from cyclic ethers and acetals by the reaction with alkylperoxy- lambda(3)-iodane.
Addition to electron deficient olefins of alpha-oxy carbon- centered radicals, generated from cyclic ethers and acetals by the reaction with alkylperoxy- lambda(3)-iodane.
Molecules (Basel, Switzerland) (2007-11-17)
Takuya Sueda, Yasunori Takeuchi, Takashi Suefuji, Masahito Ochiai
ABSTRACT
Thermal decomposition of 1-tert-butylperoxy-1,2-benziodoxol-3(1H)-one in cyclic ethers and acetals at 50 degrees C generates alpha-oxy carbon-centered radicals, which undergo an addition reaction with vinyl sulfones and unsaturated esters.
Sigma-Aldrich
Phenyl vinyl sulfone, 99%