  • Search Results
  • Sortase inhibitor phenyl vinyl sulfone inhibits Renibacterium salmoninarum adherence and invasion of host cells.
Sortase inhibitor phenyl vinyl sulfone inhibits Renibacterium salmoninarum adherence and invasion of host cells.
Diseases of aquatic organisms (2008-02-22)
Ponnerassery S Sudheesh, Samuel Crane, Kenneth D Cain, Mark S Strom
ABSTRACT
Renibacterium salmoninarum, the causative agent of bacterial kidney disease in salmonid fishes, is a Gram-positive diplococcobacillus belonging to the family Micrococcaceae. Analysis of the genome sequence of the bacterium demonstrated the presence of a sortase homolog (srtD), a gene specifying an enzyme found in Gram-positive bacteria and required for covalent anchoring of cell surface proteins. Interference of sortase activity is being examined as a target for therapeutic prevention of infection by several pathogenic Gram-positive bacterial species. In silico analysis identified 8 open reading frames containing sortase recognition motifs, suggesting these proteins are translocated to the bacterial cell wall. The sortase and potential sortase substrate genes are transcribed in R. salmoninarum, suggesting they encode functional proteins. Treatment of R. salmoninarum with phenyl vinyl sulfone (PVS) significantly reduced bacterial adherence to Chinook salmon fibronectin. In addition, the ability of the PVS-treated bacteria to adhere to Chinook salmon embryo cells (CHSE-214) in vitro was dramatically reduced compared to that of untreated bacteria. More importantly, PVS-treated bacteria were unable to invade and replicate within CHSE-214 cells (demonstrated by an intracellular growth assay and by light microscopy). When treated with PVS, R. salmoninarum was not cytopathic to CHSE-214 cells, whereas untreated bacteria produced cytopathology within a few days. These findings clearly show that PVS, a small molecule drug and a known sortase inhibitor, can interfere with the ability of R. salmoninarum to adhere and colonize fish cells, with a corresponding decrease in virulence.
Sigma-Aldrich
Phenyl vinyl sulfone, 99%