  • Search Results
  • Cognitive dietary restraint: impact on bone, menstrual and metabolic status in young women.
Cognitive dietary restraint: impact on bone, menstrual and metabolic status in young women.
Physiology & behavior (2008-05-30)
Jason D Vescovi, Jennifer L Scheid, Rayisa Hontscharuk, Mary Jane De Souza
ABSTRACT
We compared bone mineral density (BMD) and content (BMC), menstrual and metabolic status between physically active women with 1) high cognitive dietary restraint (High-CDR) (score > or = 9, n=38) and Normal-CDR (score<9, n=46) and 2) across quartiles of CDR scores. Eighty-four physically active (500+/-35 min wk(-1)) premenopausal women participated and were categorized according to their CDR score. Primary outcomes included, BMD, BMC, menstrual status, estrone-3-glucuronide (E1G) and pregnanediol-3-glucuronide (PdG) area under the curve (AUC). Secondary outcomes included resting energy expenditure (REE), total triiodothyronine, and ghrelin. Measures of body mass (59.2+/-1.1 vs. 58.5+/-1.0 kg) and percent body fat (24.7+/-1.2 vs. 23.7+/-0.7%) were similar between women with Normal-CDR and High-CDR, however the High-CDR group had lower total body (1.140+/-0.011 vs. 1.179+/-0.010 g cm(-2); p=0.015) and lumbar spine (1.114+/-0.019 vs. 1.223+/-0.022 g cm(-2); p=0.001) BMD. The prevalence of oligo-amenorrhea was higher in the High-CDR group and became increasingly greater across the CDR quartiles. There were no differences in metabolic characteristics between the High-CDR and Normal-CDR groups, however REE and the ratio of measured to predicted REE were lower in the fourth quartile (CDR scores > or = 13) compared to the second and third quartiles. Our results provide evidence that high CDR scores are associated with reduced lumbar spine and total body BMD in physically active premenopausal women. A greater frequency of menstrual disturbances in women with higher CDR scores likely played a role in the reduced total body and lumbar spine BMD.
Sigma-Aldrich
5β-Pregnane-3α,20α-diol glucuronide
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.