  • Search Results
  • Facilitated transport of penicillin G by bulk liquid membrane with TBP as carrier.
Facilitated transport of penicillin G by bulk liquid membrane with TBP as carrier.
Applied biochemistry and biotechnology (2008-06-26)
Zhongqi Ren, Yuanyuan Lv, Weidong Zhang
ABSTRACT
The facilitated transport of penicillin G from aqueous solutions to the stripping phase through bulk liquid membrane (BLM) containing TBP in 3% iso-octanol and n-butyl acetate was studied. Na(2)CO(3) solution was used as the stripping phase. Experiments were performed as a function of stirring rate, TBP concentration and type of diluent in the liquid membrane phase, pH, and initial penicillin G concentration in the feed phase, Na(2)CO(3) concentration in the stripping phase, etc. The results showed that the BLM process could carry out the simultaneous separation and concentration of penicillin G from dilute aqueous solutions, and arise "up-hill" effect due to the characteristic of non-equilibrium mass transfer. The diffusion of penicillin G complex in the liquid membrane phase played an important role in BLM process. The mass transfer mechanism of BLM for this system was also discussed.
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, puriss. p.a., ACS reagent