  • Search Results
  • Determination of methyl isothiocyanate in air downwind of fields treated with metam-sodium by subsurface drip irrigation.
Determination of methyl isothiocyanate in air downwind of fields treated with metam-sodium by subsurface drip irrigation.
Journal of agricultural and food chemistry (2008-08-06)
James E Woodrow, James N Seiber, James S LeNoir, Robert I Krieger
ABSTRACT
Air concentrations of methyl isothiocyanate (MITC) were determined near two fields treated with metam-sodium (MS) by subsurface drip irrigation. The two study fields showed measurable airborne MITC residues during application of MS and for periods up to 48 h postapplication. Using a Gaussian plume dispersion model, flux values were estimated for all of the sampling periods. On the basis of the flux estimates, the amount of MITC that volatilized within the 48 h period was about 1.4% of the applied material. Compared to other studies, MITC residues in air measured during application by subsurface drip irrigation were up to four orders-of-magnitude lower than those previously published for applications involving delivery through surface irrigation water. Our measured concentrations of MITC in field air were at levels below current regulatory guidance and thresholds for adverse human health effects.
Sigma-Aldrich
Methyl isothiocyanate, 97%
Supelco
Metam-sodium hydrate, PESTANAL®, analytical standard
Supelco
Methyl isothiocyanate, PESTANAL®, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.