  • Search Results
  • Simulation of repeated dose kinetics of methyl isobutyl ketone in humans from experimental single-dose inhalation exposure.
Simulation of repeated dose kinetics of methyl isobutyl ketone in humans from experimental single-dose inhalation exposure.
Regulatory toxicology and pharmacology : RTP (2008-09-16)
Shakil A Saghir, David L Rick
ABSTRACT
Methyl isobutyl ketone (MIBK) is a solvent used in numerous products and processes and may be present in the air of the workplace as a vapor. The American Conference of Governmental Industrial Hygienists (ACGIH) threshold limit value-time-weighted average (TLV-TWA) and TLV-short term exposure limit (TLV-STEL) for MIBK are 50 and 75 ppm, respectively. These workplace air concentration limits were set to protect workers from irritation, neurasthenic symptoms and possible adverse effects to their livers and kidneys. A recent revision of the ACGIH limit value has been proposed, to reduce the current TLV-TWA to 30 ppm. This article predicts the kinetics and accumulation of MIBK in humans exposed repeatedly in various exposure scenarios (8, 12, and 24h/day for 7 days) to the current ACGIH TLV-TWA of 50 ppm. The kinetic parameters of the model were derived from published human time-course blood MIBK data from a single 2h inhalation exposure to 48.9 ppm MIBK. The model correctly simulated single exposure experimental data with a rapid rise in blood concentration to 1.06 microg/ml within 1h and approached >or=99% steady-state blood level in 4h of exposure. MIBK was predicted to be rapidly eliminated from blood after terminating the exposure, reaching 0.53 microg/ml and 0.13 microg/ml within 0.5 and 2h post-exposure, respectively. Within 4h after the termination of exposure, blood concentration would be expected to <1% of the steady-state concentration. On the basis of these results, it is concluded that accumulation of MIBK in workers due to repeated inhalation exposure is not likely to occur at the current TLV-TWA concentration of 50 ppm.
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
2-Hexanone, analytical standard
Supelco
2-Hexanone, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.