  • Search Results
  • The effect of exercise and estrogen on osteoprotegerin in premenopausal women.
The effect of exercise and estrogen on osteoprotegerin in premenopausal women.
Bone (2008-10-22)
Sarah L West, Jennifer L Scheid, Mary Jane De Souza
ABSTRACT
The benefits of exercise are widely recognized, however physically active women can develop exercise associated menstrual cycle disturbances such as amenorrhea (i.e., estrogen deficiency) secondary to a chronic energy deficiency. To assess the effects of exercise status and estrogen deficiency on osteoprotegerin (OPG) and its relationship to bone resorption in premenopausal exercising women. Cross-sectional study of serum OPG, urinary c-telopeptides (uCTX), urinary estrone 3-glucuronide (E1G), urinary pregnanediol 3-glucuronide (PdG) and bone mineral density (BMD) measured on multiple occasions in 67 women. Volunteers were retrospectively grouped: 1) sedentary menstruating group (SedMen n=8), 2) exercising menstruating group (ExMen, n=36), and 3) exercising amenorrheic group (ExAmen, n=23). One-way ANOVAs were performed, and LSD post-hoc tests were performed when differences were detected. Subjects were similar with respect to age (24.2+/-1.0 years), weight (57.8+/-1.7 kg), and height (164.3+/-1.3 cm) (p>0.05). ExMen and ExAmen groups were more aerobically fit (p=0.003) and had less body fat (p=0.002) than the SedMen group. Resting energy expenditure/fat free mass was lowest (p=0.001) in the ExAmen groups. Mean E1G across the measurement period (p<0.001) and overall E1G exposure as assessed by E1G area under the curve (AUC) (p<0.001) were lower in the ExAmen group vs. the SedMen and ExMen groups. U-CTX-I was elevated (p=0.033) in the ExAmen group (281.8+/-40.3 microg/L/mmCr), compared with the SedMen and ExMen groups (184.5+/-22.4, 197.2+/-14.7 microg/L/mmCr, respectively). OPG was suppressed (p=0.005) in the ExAmen group (4.6+/-0.2 pmol/L) vs. ExMen group (5.2+/-0.2 pmol/L), and OPG was lower in the SedMen group (4.1+/-0.3 pmol/L) compared with the ExMen group. Findings were translated to BMD; the ExAmen group had suppressed total body BMD (p=0.014) and L2-L4 BMD (p=0.015) vs. the ExMen group. Our results suggest that OPG responds to the bone loading effect of exercise, and that suppressed OPG may play a role in the etiology of increased bone resorption observed in exercising women with chronic estrogen deficiency secondary to hypothalamic amenorrhea.
Sigma-Aldrich
5β-Pregnane-3α,20α-diol glucuronide
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.