  • Search Results
  • Signal transduction of hyaluronic acid-peptide conjugate for formyl peptide receptor like 1 receptor.
Signal transduction of hyaluronic acid-peptide conjugate for formyl peptide receptor like 1 receptor.
Bioconjugate chemistry (2008-11-15)
Eun Ju Oh, Jung-Wook Kim, Ji-Hyun Kong, Sung Ho Ryu, Sei Kwang Hahn
ABSTRACT
Agonistic and antagonistic peptides for formyl peptide receptor like 1 (FPRL1) receptor have been investigated as novel drug candidates for inflammatory diseases such as sepsis, asthma, and rheumatoid arthritis. In this work, a novel protocol for the synthesis of hyaluronic acid (HA)-peptide (CWRYMVm) conjugate for FPRL1 receptor was successfully developed for further clinical applications of peptide drugs. Aminoethyl methacrylated HA (HAAEMA) was synthesized by the coupling reaction of tetrabutyl ammonium salt of HA (HA-TBA) and AEMA using benzotriazol-1-yloxy-tris(dimethylamino) phosphonium hexafluorophosphate (BOP) in dimethyl sulfoxide (DMSO). Then, HA-AEMA was conjugated with CWRYMVm in water via Michael addition reaction between methacrylate group of HA-AEMA and thiol group in cysteine. The formation of HA-peptide conjugate was confirmed by 1H NMR and gel permeation chromatography (GPC). The average number of conjugated peptide molecules could be controlled from 5 to 23 per single HA chain. The HA-peptide conjugate showed serum stability longer than four days. In Vitro signal transduction activity of the HA-peptide conjugate for FPRL1 receptor was confirmed from the elevated levels of phospho-extracellular signal-regulated kinase (pERK) and calcium ion in FPRL1 overexpressing RBL-2H3 cells. The partially decreased biological activity of HA-peptide conjugates by the steric hindrance of HA was recovered after its degradation by hyaluronidase treatment.
Sigma-Aldrich
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate, 97%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade