  • Search Results
  • Facilitated acquisition of the classically conditioned eyeblink response in women taking oral contraceptives.
Facilitated acquisition of the classically conditioned eyeblink response in women taking oral contraceptives.
Behavioural pharmacology (2008-11-21)
Kevin D Beck, Joselyn McLaughlin, Michael T Bergen, Tara P Cominski, Roberta L Moldow, Richard J Servatius
ABSTRACT
Although anecdotal reports suggest that associative learning processes are affected by menstrual phase, empirical evidence has been equivocal. Moreover, there is a dearth of research concerning fluctuations of artificial or exogenous female hormones on learning and memory. Therefore, in this preliminary study we assessed learning in women who take oral contraceptives and those who do not during the three phases of the menstrual cycle: early, middle, and later cycle. The behavioral assessment included short-trace eyeblink conditioning, acoustic startle reactivity, and a fine motor coordination task (grooved pegboard). Oral contraceptive users generally acquired the conditioned eyeblink response better than non-users. Similar enhancements were observed for fine motor coordination and startle responsiveness. Further research will need to distinguish whether the hormone influence is upon the associative processes or the sensory-motor pathways involved in nonassociative learning.
Sigma-Aldrich
5β-Pregnane-3α,20α-diol glucuronide
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.