  • Search Results
  • Calcium fingerprints induced by calmodulin interactors in eukaryotic cells.
Calcium fingerprints induced by calmodulin interactors in eukaryotic cells.
Biochimica et biophysica acta (2009-01-10)
Rania Dagher, Christian Brière, Marie Fève, Maria Zeniou, Claire Pigault, Christian Mazars, Hervé Chneiweiss, Raoul Ranjeva, Marie-Claude Kilhoffer, Jacques Haiech
ABSTRACT
Calcium (Ca2+) is a ubiquitous second messenger which promotes cell responses through transient changes in intracellular concentrations. The prominent role of Ca2+ in cell physiology is mediated by a whole set of proteins constituting a Ca2+-signalling toolkit involved in Ca2+-signal generation, deciphering and arrest. The different Ca2+-signalosomes deliver Ca2+-signals with spatial and temporal dynamics to control the function of specific cell types. Among the intracellular proteins involved in Ca2+-signal deciphering, calmodulin (CaM) plays a pivotal role in controlling Ca2+-homeostasis and downstream Ca2+-based signalling events. Due to its ubiquitous expression in eukaryotic cells and the variety of proteins it interacts with, CaM is central in Ca2+-signalling networks. For these reasons, it is expected that disrupting or modifying CaM interactions with its target proteins will affect Ca2+-homeostasis and cellular responses. The resulting calcium response will vary depending on which interactions between CaM and target proteins are altered by the molecules and on the specific Ca2+-toolkit expressed in a given cell, even in the resting state. In the present paper, the effect of six classical CaM interactors (W5, W7, W12, W13, bifonazole and calmidazolium) was studied on Ca2+-signalling in tumor initiating cells isolated from human glioblastoma (TG1) and tobacco cells (BY-2) using the fluorescent Ca2+-sensitive Indo-1 dye and aequorin, respectively. Various Ca2+-fingerprints were obtained depending both on the CaM interactor used and the cell type investigated. These data demonstrate that interaction between the antagonists and CaM results in a differential inhibition of CaM-dependent proteins involved in Ca2+-signal regulation. In addition, the distinct Ca2+-fingerprints in tobacco and human tumor initiating glioblastoma cells induced by a given CaM interactor highlight the specificity of the Ca2+-signalosome in eukaryotic cells.
Product Description
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.