  • Search Results
  • Water-immiscible solvents as diluents in reversed-phase liquid chromatography.
Water-immiscible solvents as diluents in reversed-phase liquid chromatography.
Journal of chromatography. A (2009-02-25)
Eric Loeser, Stanislaw Babiak, Patrick Drumm
ABSTRACT
It has recently been shown that the use of strong organic solvents as diluent is possible in RPLC, provided that the solvent used as diluent is retained more strongly by the column than the analytes in the sample. In this study, the phenomenon was further studied experimentally using several water-immiscible solvents (ethyl acetate, isopropyl acetate, and methyl isobutyl ketone) and several model analyte compounds. In all cases, analyte peak distortion was minimal provided the analyte eluted earlier than the diluting solvent, in contrast to substantial broadening and distortion when the analyte eluted after the diluting solvent. The potential analytical utility of this approach is discussed, and an example of a practical application is also presented.
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Sigma-Aldrich
Isopropyl acetate, ≥99.6%
Sigma-Aldrich
Isopropyl acetate, puriss. p.a., ≥99.5% (GC)
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Supelco
Isopropyl acetate, analytical standard
Sigma-Aldrich
Isopropyl acetate, 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Sigma-Aldrich
Isopropyl acetate, ≥99%, FCC
Supelco
2-Hexanone, analytical standard
Supelco
2-Hexanone, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.