Capillary electrophoresis for screening of 20S proteasome inhibitors.
Analytical biochemistry (2009-07-21)
Wenjing Chen, Ke Mou, Bo Xu, Xiaomei Ling, Jingrong Cui, Ping Xu
ABSTRACT
A method for studying 20S proteasome inhibitors by capillary electrophoresis (CE) has been developed. Proteasome plays a fundamental role in degrading key regulatory proteins. The 20S proteasome can degrade intrinsically disordered proteins in an ATP-independent manner without additional "helper" molecules. The discovery of new proteasome inhibitors with little or no toxicity is highly desirable in anticancer therapy. In this study, the inhibitory effects of MG132 and MG115 on the 20S proteasome were evaluated by CE for the first time. The optimized CE conditions were as follows: fused-silica capillary of 30 cm effective length and 75 microm internal diameter, pressure injection of 0.5 psi for 5 s, 50 mM Hepes buffer (pH 7.6) with 2% dimethyl sulfoxide, constant voltage of 20 kV, and detection wavelength at 340 nm. Also, the new method was used to study the inhibitory effects of 30 novel peptidyl vinyl ester derivatives of MG132. The 50% inhibition concentrations (IC(50) values) of MG132 and MG115 were 40.0 and 84.7 nM, respectively. Two new compounds, XP32 and XP35, showed considerable inhibitory effects on the 20S proteasome. When the concentrations of them were fixed at 172 nM, their inhibition rates were 36.2% and 29.1%, respectively. The results showed that the CE method was powerful, sensitive, and fast and required little sample. It could be employed as one of the reliable drug screening methods for 20S proteasome inhibitors.
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.