  • Search Results
  • Determination of cyanide, in urine and gastric content, by electrospray ionization tandem mass spectrometry after direct flow injection of dicyanogold.
Determination of cyanide, in urine and gastric content, by electrospray ionization tandem mass spectrometry after direct flow injection of dicyanogold.
Analytica chimica acta (2009-09-08)
Kayoko Minakata, Hideki Nozawa, Kunio Gonmori, Masako Suzuki, Osamu Suzuki
ABSTRACT
A rapid and sensitive electrospray ionization tandem mass spectrometric (ESI-MS-MS) procedure was developed for the determination of cyanide (CN(-)). CN(-) in biological fluids was reacted with NaAuCl(4) to produce dicyanogold, Au(CN)(2)(-), which was extracted with methyl isobutyl ketone (MIBK). One microliter of the extract was injected directly into an ESI-MS-MS instrument. Quantification of CN(-) was performed by selected reaction monitoring of the product ion CN(-) at m/z 26 that derived from precursor ion Au(CN)(2)(-) at m/z 249. CN(-) could be measured in the quantification range of 10(-7) to 5x10(-5) M with the limit of detection at 4x10(-8) M using 10 microL of urine within 10 min. A victim's urine and gastric content were diluted with water to 4-fold and 500-fold and measured, respectively.
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Sigma-Aldrich
Potassium dicyanoaurate(I), 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
2-Hexanone, analytical standard
Sigma-Aldrich
Potassium dicyanoaurate(I), 99.95% trace metals basis
Supelco
2-Hexanone, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.