  • Search Results
  • Comparision of the volatile components of unripe and ripe Japanese apricot (Prunus mume Sieb. et Zucc.).
Comparision of the volatile components of unripe and ripe Japanese apricot (Prunus mume Sieb. et Zucc.).
Natural product research (2009-10-24)
Mitsuo Miyazawa, Nozomu Shirakawa, Hirotoshi Utsunomiya, Ken-ichi Inada, Tomoki Yamada
ABSTRACT
The composition of the volatile components from unripe (I) and ripe (II) Japanese apricot (Prunus mume Sieb. et Zucc.) have been investigated. Seventy-six volatile components were identified, 25 for the first time as volatile constituents of Japanese apricot. The main components were benzaldehyde (I, 59.16%; II, 1.81%), isolongifololyl acetate (II, 19.21%), palmitic acid (I, trace; 10.22%), linalool (I, 9.93%; II, 7.34%), and butyl acetate (II, 8.30%). Unripe Japanese apricot have a green colour due to the main components being aldehydes. On the contrary, ripe Japanese apricot had a fruity note due to the increase of the ratio of esters during ripening.
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, puriss. p.a., ACS reagent