  • Search Results
  • Endostatin expression in neurons during the early stage of cerebral ischemia is associated with neuronal apoptotic cell death in adult hypertensive rat model of stroke.
Endostatin expression in neurons during the early stage of cerebral ischemia is associated with neuronal apoptotic cell death in adult hypertensive rat model of stroke.
Brain research (2009-11-28)
Qinghua Hou, Li Ling, Fang Wang, Shihui Xing, Zhong Pei, Jinsheng Zeng
ABSTRACT
Endostatin (ES) has been recognized as a potent anti-angiogenic factor. We here investigated the expression of ES in ischemic brain and the consequence of cells expressing ES after stroke in adult stroke-prone renovascular hypertensive rats. A single dose of Ca-074ME, a membrane-permeable cathepsin B (CB) specific inhibitor, or vehicle was given by intraperitoneal injection immediately after distal middle cerebral artery occlusion (dMCAO), ES expression was evaluated using fluorescent immunohistochemistry staining, and CB enzyme activity was tested by measuring the free 7-amino-4-methylcoumarin (AMC) released by CB from its' specific substrate, the Z-Arg-Arg-7-amido-4-methylcoumarin. ES immunoreactivity (IR) was significantly up-regulated as early as 6 h and returned to baseline level at 3 days in peri-infarct area following dMCAO. Double-staining experiment revealed that the majority of ischemia-induced ES positive cells were neurons. Furthermore, ES was co-labeled with CB and Cleaved Caspase-3(Asp175) whereas treatment with Ca-074ME reduced up-regulation of ES expression and attenuated apoptosis in peri-infarct neurons. Collectively, our data suggest that peri-infarct neurons express ES during the early stage of cerebral ischemia and treatment with Ca-074ME attenuates ES expression and apoptosis in peri-infarct neurons.
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.