  • Search Results
  • Fluorescence lifetime tuning--a novel approach to study flip-flop kinetics in supported phospholipid bilayers.
Fluorescence lifetime tuning--a novel approach to study flip-flop kinetics in supported phospholipid bilayers.
Journal of fluorescence (2009-12-30)
Anna Kułakowska, Piotr Jurkiewicz, Jan Sýkora, Ales Benda, Yves Mely, Martin Hof
ABSTRACT
In the present work we introduce a straightforward fluorescent assay that can be applied in studies of the transbilayer movement (flip-flop) of fluorescent lipid analogues across supported phospholipid bilayers (SPBs). The assay is based on the distance dependent fluorescence quenching by light absorbing surfaces. Applied to SPBs this effect leads to strong differences in fluorescence lifetimes when the dye moves from the outer lipid leaflet to the leaflet in contact with the support. Herein, we present the basic principles of this novel approach, and comment on its advantages over the commonly used methods for investigating flip-flop dynamics across lipid bilayers. We test the assay on the fluorescent lipid analog Atto633-DOPE and the 3-hydroxyflavone F2N12S probe in SPBs composed of DOPC/ DOPS lipids. Moreover, we compare and discuss the flip-flop rates of the probes with respect to their lateral diffusion coefficients.
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Sigma-Aldrich
Atto 488, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 488-Biotin, BioReagent, suitable for fluorescence, ~75% (HPCE)
Sigma-Aldrich
Atto 488 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 633, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 488 amine
Sigma-Aldrich
Atto 488 azide
Supelco
Atto 633 NHS ester, BioReagent, suitable for fluorescence, ≥90.0% (coupling to amines)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium salt, ~95%
Sigma-Aldrich
Atto 488 iodoacetamide
Sigma-Aldrich
Atto 633 amine, suitable for fluorescence
Sigma-Aldrich
Atto 633 iodoacetamide, suitable for fluorescence
Sigma-Aldrich
Atto 633 azide, suitable for fluorescence, ≥80.0% (HPCE)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.