MilliporeSigma
A new decalin derivative from red yeast rice.
Journal of Asian natural products research (2010-02-26)
Yun-Tian Zhang, Ying Wang, Xian-Tao Zhang, Dong-Ling Wu, Xiao-Qi Zhang, Wen-Cai Ye
ABSTRACT
A new decalin derivative, monascusic acid A (1), together with a new natural product (2), was isolated from the ethanol extract of red yeast rice. Their structures were elucidated by spectroscopic methods.
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%