  • Search Results
  • [Preparation and properties of isocitrate lyase isoforms from the cotyledons of Glycine max L].
[Preparation and properties of isocitrate lyase isoforms from the cotyledons of Glycine max L].
Prikladnaia biokhimiia i mikrobiologiia (2010-03-05)
A T Eprintsev, E V Diachenko, T V Lykova, C T H Kuen, V N Popov
ABSTRACT
A four-stage purification procedure including ammonium sulfate precipitation and ion exchange chromatography on DEAE cellulose has been elaborated for isolation of isocitrate lyase (EC 4.1.3.1) isoforms from the cotyledons of soybean Glycine max L. Electrophoretically homogeneous preparations of two forms of the enzyme with specific activity of 5.28 and 5.81 U/mg protein have been obtained. Comparison of physicochemical, kinetic, and regulation characteristics of the isoforms obtained revealed fundamental differences between them. Thus, the isoform that migrated quickly in PAAG had a much lower affinity to isocitrate (K(M) - 50 microM) than the slowly migrating form (K(M) - 16 microM). It has been shown that the conservation of activity of the isoforms obtained depends on the presence of divalent cations (Mn2+ and Mg2+) in the medium. It is suggested to use the isoforms of isocitrate lyase isolated from soybeans for the development of biosensors for biochemical and kinetic assays.
Sigma-Aldrich
DL-Isocitric acid trisodium salt hydrate, ≥93%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.