  • Search Results
  • Electric conductivities of 1:1 electrolytes in high-temperature ethanol along the liquid-vapor coexistence curve. I. NaBr, KBr, and CsBr.
Electric conductivities of 1:1 electrolytes in high-temperature ethanol along the liquid-vapor coexistence curve. I. NaBr, KBr, and CsBr.
The Journal of chemical physics (2010-03-25)
Koji Takahata, Taka-aki Hoshina, Noriaki Tsuchihashi, Kazuyasu Ibuki, Masakatsu Ueno
ABSTRACT
The molar electric conductivities Lambda of NaBr, KBr, and CsBr were measured in liquid ethanol at temperatures from 60 to 220 degrees C along the liquid-vapor coexistence curve. The limiting molar electrolyte conductivities Lambda(o) and the molar association constants K(A) were determined by the analysis of the concentration dependence of Lambda. The friction coefficients zeta for the Na(+), K(+), Cs(+), and Br(-) ions were estimated from Lambda(o) by an assumption that the cationic transference number of KBr is independent of temperature and density. The density dependences of zeta thus obtained together with literature values at higher densities (lower temperatures) were examined. zeta increases with decreasing density at densities above 2.0rho(c), where rho(c)=0.276 g cm(-3) is the critical density. At lower densities, the density dependences of zeta depend on ion. The relative contribution of the nonviscous effect in zeta was estimated by Deltazeta/zeta, where Deltazeta was the difference between zeta and the Stokes friction coefficient. At densities above 2.7rho(c), Deltazeta/zeta slightly decreases with decreasing density except for the Cs(+) ion. At densities below 2.7rho(c), Deltazeta/zeta increases with decreasing density and the density dependence is larger for larger ion. The results at densities above 2.2rho(c) were well explained by the Hubbard-Onsager (HO) dielectric friction theory [J. Hubbard, J. Chem. Phys. 68, 1649 (1978)] based on the sphere-in-continuum model. Below 2.2rho(c), however, experimental Deltazeta/zeta tends to be larger than the prediction of the HO theory. The lower limit density of the validity range of the HO theory is slightly higher in ethanol than in methanol.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Cesium bromide, 99.9% trace metals basis
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Sodium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Cesium bromide, 99.999% trace metals basis
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%