  • Search Results
  • Effect of oral sebacic Acid on postprandial glycemia, insulinemia, and glucose rate of appearance in type 2 diabetes.
Effect of oral sebacic Acid on postprandial glycemia, insulinemia, and glucose rate of appearance in type 2 diabetes.
Diabetes care (2010-08-21)
Amerigo Iaconelli, Amalia Gastaldelli, Chiara Chiellini, Donatella Gniuli, Angela Favuzzi, Christophe Binnert, Katherine Macé, Geltrude Mingrone
ABSTRACT
Dicarboxylic acids are natural products with the potential of being an alternate dietary source of energy. We aimed to evaluate the effect of sebacic acid (a 10-carbon dicarboxylic acid; C10) ingestion on postprandial glycemia and glucose rate of appearance (Ra) in healthy and type 2 diabetic subjects. Furthermore, the effect of C10 on insulin-mediated glucose uptake and on GLUT4 expression was assessed in L6 muscle cells in vitro. Subjects ingested a mixed meal (50% carbohydrates, 15% proteins, and 35% lipids) containing 0 g (control) or 10 g C10 in addition to the meal or 23 g C10 as a substitute of fats. In type 2 diabetic subjects, the incremental glucose area under the curve (AUC) decreased by 42% (P<0.05) and 70% (P<0.05) in the 10 g C10 and 23 g C10 groups, respectively. At the largest amounts used, C10 reduced the glucose AUC in healthy volunteers also. When fats were substituted with 23 g C10, AUC of Ra was significantly reduced on the order of 18% (P<0.05) in both healthy and diabetic subjects. The insulin-dependent glucose uptake by L6 cells was increased in the presence of C10 (38.7±10.3 vs. 11.4±5.4%; P=0.026). This increase was associated with a 1.7-fold raise of GLUT4. Sebacic acid significantly reduced hyperglycemia after a meal in type 2 diabetic subjects. This beneficial effect was associated with a reduction in glucose Ra, probably due to lowered hepatic glucose output and increased peripheral glucose disposal.
Sigma-Aldrich
Sebacic acid, 99%
Sigma-Aldrich
Sebacic acid, ≥95.0% (GC)