  • Search Results
  • Biological evaluation of the inhibition of neutrophil elastase by a synthetic beta-lactam derivative.
Biological evaluation of the inhibition of neutrophil elastase by a synthetic beta-lactam derivative.
European journal of cell biology (1990-08-01)
J L Maillard, C Favreau, M Reboud-Ravaux, R Kobaiter, R Joyeau, M Wakselman
ABSTRACT
A novel beta-lactam derivative, N-(2-chloromethylphenyl) 3,3-difluoroazetidin-2-one, which behaves as a time-dependent inactivator of leukocyte elastase, has been tested in biological models designed to detect its potential therapeutic value in the treatment of emphysema. Its effect on two types of leukocyte elastase, purified human leukocyte elastase and elastase freshly discharged upon stimulation of guinea pig polymorphonuclear neutrophils, was examined using three methods: the cleavage of a chromogenic peptide substrate, MeO-Suc-Ala-Ala-Pro-Val-NA, the lysis and solubilization of tritiated elastin and the microscopic examination of the damage to lung elastic network. The inhibitor was shown to be effective at preventing proteolysis due to leukocyte elastase. Besides its low cellular toxicity, no apparent hindrance of its efficiency was found in the above quasi in vivo environment. This suggests that this inhibitor may be of potential therapeutic value in elastase-related pathology.
Sigma-Aldrich
N-Methoxysuccinyl-Ala-Ala-Pro-Val p-nitroanilide, elastase substrate
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.