Chiral supramolecular self-assembly of rubrene.
Physical chemistry chemical physics : PCCP (2010-09-08)
Li Wang, Huihui Kong, Xing Song, Xiaoqing Liu, Hongming Wang
ABSTRACT
Large area chiral supramolecular self-assembly of rubrene has been achieved on Au(111) surface at room temperature. The basic building block of such self-assembled layers consists of two double-layered Y-shape supramolecular structures containing eight twisted rubrene molecules. Chirality is maintained and transferred from the bottom layer to the top layer in the successive molecular layers for up to five layers. Such chiral multilayers can be potential candidates in enantioselective catalysis and chiral separations.
Sigma-Aldrich
Rubrene, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Rubrene, ≥98%
Sigma-Aldrich
Rubrene, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.