  • Search Results
  • Use of a holder-vacuum tube device to save on-site hands in preparing urine samples for head-space gas-chromatography, and its application to determine the time allowance for sample sealing.
Use of a holder-vacuum tube device to save on-site hands in preparing urine samples for head-space gas-chromatography, and its application to determine the time allowance for sample sealing.
Industrial health (2010-09-09)
Toshio Kawai, Kimiaki Sumino, Fumiko Ohashi, Masayuki Ikeda
ABSTRACT
To facilitate urine sample preparation prior to head-space gas-chromatographic (HS-GC) analysis. Urine samples containing one of the five solvents (acetone, methanol, methyl ethyl ketone, methyl isobutyl ketone and toluene) at the levels of biological exposure limits were aspirated into a vacuum tube via holder, a device commercially available for venous blood collection (the vacuum tube method). The urine sample, 5 ml, was quantitatively transferred to a 20-ml head-space vial prior to HS-GC analysis. The loaded tubes were stored at +4 ℃ in dark for up to 3 d. The vacuum tube method facilitated on-site procedures of urine sample preparation for HS-GC with no significant loss of solvents in the sample and no need of skilled hands, whereas on-site sample preparation time was significantly reduced. Furthermore, no loss of solvents was detected during the 3-d storage, irrespective of hydrophilic (acetone) or lipophilic solvent (toluene). In a pilot application, high performance of the vacuum tube method in sealing a sample in an air-tight space succeeded to confirm that no solvent will be lost when sealing is completed within 5 min after urine voiding, and that the allowance time is as long as 30 min in case of toluene in urine. The use of the holder-vacuum tube device not only saves hands for transfer of the sample to air-tight space, but facilitates sample storage prior to HS-GC analysis.
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
2-Hexanone, analytical standard
Supelco
2-Hexanone, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.