  • Search Results
  • Direct electrospray ionization mass spectrometry quantitative analysis of sebacic and terephthalic acids in biodegradable polymers.
Direct electrospray ionization mass spectrometry quantitative analysis of sebacic and terephthalic acids in biodegradable polymers.
Analytical chemistry (2011-01-06)
Paola Rizzarelli, Daniela Zampino, Loredana Ferreri, Giuseppe Impallomeni
ABSTRACT
A direct, rapid, and easy electrospray ionization mass spectrometry (ESI-MS) method to determine concentrations of sebacic acid (SA) and terephthalic acid (TA) residues in biodegradable copolymers was developed. Copolyester samples were synthesized from 1,4-butanediol and sebacic and terephthalic acids by melt polymerization. Extraction of monomers was performed in methanol. Their concentrations were determined by direct infusion ESI-MS, without chromatographic separation, using 1,12-dodecanedioic acid (DDA) as an internal standard. Calibration curves were obtained by plotting the ratio of the areas of the peaks relative to monomers and DDA standard as a function of their concentration ratio. We validated the method by determining the concentration of TA residue using both the ESI-MS protocol and high-performance liquid chromatography (HPLC) analysis with UV detection. The linearity range and the detection limit of this assay were 0.1-5.0 and 0.01 ppm for SA and 0.1-6.0 and 0.03 ppm for TA. This assay represents a useful alternative to conventional methods currently employed for acid quantification, resulting advantageous for its speed and high sensitivity.
Sigma-Aldrich
Sebacic acid, 99%
Sigma-Aldrich
Sebacic acid, ≥95.0% (GC)