  • Search Results
  • [Study on the chemical constituents from the bulbs of Zephyranthes candida].
[Study on the chemical constituents from the bulbs of Zephyranthes candida].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-03-26)
Jian-Sai Yang, Xu Feng, Yu Chen, Xing-Zeng Zhao, Ming Wang, Qi-Zhi Wang, Yun-Fa Dong
ABSTRACT
To study the chemical constituents from the bulbs of Zephyranthes candida. The chemical constituents were separated and purified by means of several chromatographies and their structures were elucidated by spectroscopic methods. Six compounds were isolated and identified as trisphaeridine (I), p-hydroxybenzaldehyde (II), sucrose (III), (-)-amarbellisine (IV), lycorine (V), (+)-haemanthamine (VI). Compounds I - IV are obtained from this genus for the first time.
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
Lycorine hydrochloride, ≥98% (TLC), powder