  • Search Results
  • The applicability of the PTTL dose re-analysis method to the Harshaw LiF:Mg,Cu,P material.
The applicability of the PTTL dose re-analysis method to the Harshaw LiF:Mg,Cu,P material.
Radiation protection dosimetry (2011-04-01)
M Moscovitch, L Benevides, A Romanyukha, F Hull, M Duffy, S Voss, K J Velbeck, I Nita, J E Rotunda
ABSTRACT
The phototransferred thermoluminescence (PTTL) technique is applied to the Harshaw LiF:Mg,Cu,P material. It is demonstrated that using 254-nm UV light, dose levels as low as 0.2 mGy can be re-estimated. The PTTL efficiency was found to be ∼ 6 % in the dose range of 0.2 mGy-1 Gy, and it appears to be dose-independent. This implies that a simple calibration factor could be applied to the PTTL data for the re-estimation of dose levels. It was demonstrated that with a proper choice of the TL readout parameters, and the UV-light irradiation conditions, dose levels that are relevant to personal or environmental dosimetry can be re-estimated.
Sigma-Aldrich
Lithium fluoride, powder, -300 mesh
Sigma-Aldrich
Lithium fluoride, powder, <100 μm, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Lithium fluoride, Precipitated, 99.995%
Sigma-Aldrich
Lithium-6Li fluoride, 95 atom % 6Li, 99% (CP)