  • Search Results
  • The enantiomers of etodolac, a racemic anti-inflammatory agent, play different roles in efficacy and gastrointestinal safety.
The enantiomers of etodolac, a racemic anti-inflammatory agent, play different roles in efficacy and gastrointestinal safety.
Biological & pharmaceutical bulletin (2011-05-03)
Naoki Inoue, Masaki Nogawa, Sunao Ito, Koyuki Tajima, Sato Kume, Takashi Kyoi
ABSTRACT
The anti-inflammatory agent etodolac is used worldwide and it has a good gastrointestinal safety profile. Etodolac consists of two enantiomers, S- and R-etodolac. Here, we investigated the beneficial activities of racemic etodolac and its enantiomers. First, we compared S- and R-etodolac in terms of their inhibition of cyclooxygenase (COX) activity in vitro and their suppression of paw swelling in adjuvant-induced arthritic rats. The COX-2 inhibitory and anti-inflammatory effects of etodolac were found to be due to the S-enantiomer. We previously reported that etodolac attenuates allodynia in a mouse model of neuropathic pain by a COX-2-independent mechanism [N. Inoue et al., J. Pharmacol. Sci., 109, 600-605 (2009)]. In the present study, we showed that the anti-allodynic effects of etodolac in mice were also due to the S-enantiomer. In addition, we investigated the ulcerogenic activity of racemic etodolac and its enantiomers. At high doses, racemic etodolac showed a lower gastric lesion index in rats than the equivalent dose of S-etodolac. In contrast, R-etodolac showed no ulcerogenic activity and even showed protection against HCl/ethanol-induced gastric damage in rats. In conclusion, S-etodolac exhibited anti-inflammatory effects mediated by COX-2 inhibition and anti-allodynic effects that were independent of COX-2 inhibition, while R-etodolac showed gastroprotective effects that may contribute to the low gastrointestinal toxicity of racemic etodolac. Our results show that each enantiomer plays a different role in the efficacy and gastrointestinal safety of etodolac.
Sigma-Aldrich
Etodolac
Etodolac for peak identification, European Pharmacopoeia (EP) Reference Standard
Etodolac, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.