  • Search Results
  • A novel and unusually long-lived chemiluminophore based on the 7-hydroxycoumarin scaffold.
A novel and unusually long-lived chemiluminophore based on the 7-hydroxycoumarin scaffold.
Chemical communications (Cambridge, England) (2011-05-17)
Jan Kočí, Virgile Grandclaude, Marc Massonneau, Jean-Alexandre Richard, Anthony Romieu, Pierre-Yves Renard
ABSTRACT
Synthesis and chemiluminescent properties of a new 1,2-dioxetane chemiluminophore bearing a 7-hydroxycoumarin moiety are presented. The 1,2-dioxetane decomposition ended up with strong and long-lived emission of light. This new structure opens way to the development of a new generation of bright chemiluminescent bio-probes.
Sigma-Aldrich
Umbelliferone, 99%
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)