  • Search Results
  • Chemical bonding in copper-based transparent conducting oxides: CuMO2 (M = In, Ga, Sc).
Chemical bonding in copper-based transparent conducting oxides: CuMO2 (M = In, Ga, Sc).
Journal of physics. Condensed matter : an Institute of Physics journal (2011-08-05)
K G Godinho, B J Morgan, J P Allen, D O Scanlon, G W Watson
ABSTRACT
The geometry and electronic structure of copper-based p-type delafossite transparent conducting oxides, CuMO(2) (M = In, Ga, Sc), are studied using the generalized gradient approximation (GGA) corrected for on-site Coulomb interactions (GGA + U). The bonding and valence band compositions of these materials are investigated, and the origins of changes in the valence band features between group 3 and group 13 cations are discussed. Analysis of the effective masses at the valence and conduction band edge explains the experimentally reported conductivity trends.
Sigma-Aldrich
Gallium(III) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Scandium(III) oxide, 99.9% trace rare earth metals basis
Sigma-Aldrich
Indium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Indium(III) oxide, nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis
Sigma-Aldrich
Indium(III) oxide, 99.998% trace metals basis
Sigma-Aldrich
Scandium(III) oxide, powder, 99.995% trace rare earth metals basis
Sigma-Aldrich
Scandium(III) oxide, 99.99% trace rare earth metals basis