  • Search Results
  • Solubilization of triphenylamine, triphenylphosphine, triphenylphosphineoxide and triphenylmethanol in single and binary surfactant systems.
Solubilization of triphenylamine, triphenylphosphine, triphenylphosphineoxide and triphenylmethanol in single and binary surfactant systems.
Journal of colloid and interface science (2011-09-13)
Mohammad Amin Mir, Oyais Ahmad Chat, Muzaffar Hussain Najar, Mohammad Younis, Aijaz Ahmad Dar, Ghulam Mohammad Rather
ABSTRACT
Solubilization and co-solubilization of triphenyls (TPs) viz., triphenylphosphine (TPP), triphenylphosphineoxide (TPPO), triphenylamine (TPA) and triphenylmethanol (TPM) were studied in various single and binary surfactant systems at 25 °C using UV-visible spectroscopy and HPLC. The solubilization capacities of different micelles towards TPs were found to be a function of the nature and structure of solubilizates, locus of solubilization, size of micelles and the nature of interactions between the solubilizate and micelles. The effect of surfactant mixing on the solubilization of TPs was evaluated using the Regular Solution Approach (RSA). The solubility enhancement of TPs within mixed micelles relative to that observed in single surfactant systems was explained in light of the structural micellar changes associated with the mixing of ionic and non-ionic surfactants. Moreover, kinetics of oxidation of TPP by hydrogen peroxide investigated in these surfactant systems was found to be sensitive to the nature of micelle and the locus of solubilization of TPP within the micelles.
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine oxide, 98%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine hydrobromide, 97%