  • Search Results
  • Electric field polarization in conventional density functional theory: from quasilinear to two-dimensional and three-dimensional extended systems.
Electric field polarization in conventional density functional theory: from quasilinear to two-dimensional and three-dimensional extended systems.
The Journal of chemical physics (2011-10-28)
Bernard Kirtman, Valentina Lacivita, Roberto Dovesi, Heribert Reis
ABSTRACT
The large overshoot in (hyper)polarizabilities of quasilinear (1D) chains calculated by applying density functional theory with conventional functionals is investigated for several 2D and 3D extended systems. These systems include arrays of molecular hydrogen chains, as well as 2D coronene-type structures and LiF in 1D, 2D, and 3D. Contrary to a recently proposed model it is found that the overshoot persists in all of these cases. A simple explanation is provided by an analysis of the field-induced charges for molecular hydrogen, which shows an excessive buildup at the chain ends regardless of where the chain is located within the 2D and 3D array.
Sigma-Aldrich
Lithium fluoride, powder, -300 mesh
Sigma-Aldrich
Coronene, 97%
Sigma-Aldrich
Lithium fluoride, powder, <100 μm, ≥99.98% trace metals basis
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Coronene, BCR®, certified reference material
Sigma-Aldrich
Lithium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Lithium fluoride, Precipitated, 99.995%
Sigma-Aldrich
Lithium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium-6Li fluoride, 95 atom % 6Li, 99% (CP)