  • Search Results
  • The effect of renal administration of a selective cyclooxygenase-2 inhibitor or stable prostaglandin I2 analog on the progression of sclerotic glomerulonephritis in rats.
The effect of renal administration of a selective cyclooxygenase-2 inhibitor or stable prostaglandin I2 analog on the progression of sclerotic glomerulonephritis in rats.
Clinical and experimental nephrology (2011-12-08)
Yukiko Nozawa, Ayako Sato, Hoglan Piao, Tetsuo Morioka, Ichiei Narita, Takashi Oite
ABSTRACT
There is increasing evidence that a change in glomerular hemodynamics may promote the development of glomerulosclerosis. In this study, we focused on the pharmacological effects of 2 contrasting agents, etodolac, a preferential cyclooxygenase-2 inhibitor, and beraprost sodium (BPS), a prostaglandin I(2) analog, delivered renally, on the disease course of progressive anti-Thy-1 (ATS) glomerulonephritis. Intravital microscopic analysis showed that the diameters of glomerular capillaries and glomerular blood flow in unilaterally nephrectomized (Nx) rats treated locally with BPS were significantly increased, as compared to those of Nx rats treated locally with normal saline (NS) or etodolac. We then examined the effects of BPS and etodolac on the course of progressive glomerulosclerosis. Mesangial cell proliferation, adhesion of glomerular capillary tufts and crescent formation in the BPS-treated group appeared to be more severe compared to the ATS + NS and the ATS + etodolac groups. Scoring of mesangial proliferation and glomerulosclerosis revealed that local BPS treatment significantly worsened glomerular pathology. At day 28, there were significant differences in blood flow between the ATS + etodolac group and both the ATS + NS and ATS + BPS groups, indicating that local treatment with etodolac enhanced the recovery of glomerular circulation. This study provides hemodynamic-based evidence showing that disturbance of intraglomerular microcirculation is a critical marker for progressive glomerulonephritis.
Sigma-Aldrich
Etodolac
Etodolac for peak identification, European Pharmacopoeia (EP) Reference Standard
Etodolac, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.