  • Search Results
  • Interaction between nuclear graphite and molten fluoride salts: a synchrotron radiation study of the substitution of graphitic hydrogen by fluoride ion.
Interaction between nuclear graphite and molten fluoride salts: a synchrotron radiation study of the substitution of graphitic hydrogen by fluoride ion.
The journal of physical chemistry. A (2012-01-19)
Xinmei Yang, Shanglei Feng, Xingtai Zhou, Hongjie Xu, T K Sham
ABSTRACT
The interaction between nuclear graphite and molten fluoride salts (46.5 mol % LiF/11.5 mol % NaF/42 mol % KF) is investigated by synchrotron X-ray diffraction and C K-edge X-ray absorption near-edge structure (XANES). It is found that there are a large number of H atoms in IG-110 nuclear graphite, which is attributed to the residual C-H bond after the graphitization process of petroleum coke and pitch binder. The elastic recoil detection analysis indicates that H atoms are uniformly distributed in IG-110 nuclear graphite, in excellent agreement with the XANES results. The XANES results indicate that the immersion in molten fluoride salts at 500 °C led to H atoms in nuclear graphite partly substituted by the fluorine from fluoride salts to form C-F bond. The implications of these findings are discussed.
Sigma-Aldrich
Lithium fluoride, powder, -300 mesh
Sigma-Aldrich
Lithium fluoride, powder, <100 μm, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Lithium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Lithium fluoride, Precipitated, 99.995%
Sigma-Aldrich
Lithium-6Li fluoride, 95 atom % 6Li, 99% (CP)