  • Search Results
  • Immobilization of lipase B from Candida antarctica on porous styrene-divinylbenzene beads improves butyl acetate synthesis.
Immobilization of lipase B from Candida antarctica on porous styrene-divinylbenzene beads improves butyl acetate synthesis.
Biotechnology progress (2012-01-25)
Natália G Graebin, Andréa B Martins, André S G Lorenzoni, Cristina Garcia-Galan, Roberto Fernandez-Lafuente, Marco A Z Ayub, Rafael C Rodrigues
ABSTRACT
A new biocatalyst of lipase B from Candida antarctica (MCI-CALB) immobilized on styrene-divinylbenzene beads (MCI GEL CHP20P) was compared with the commercial Novozym 435 (immobilized lipase) in terms of their performances as biocatalysts for the esterification of acetic acid and n-butanol. The effects of experimental conditions on reaction rates differed for each biocatalyst, showing different optimal values for water content, temperature, and substrate molar ratio. MCI-CALB could be used at higher acid concentrations, up to 0.5 M, while Novozym 435 became inactivated at these acid concentrations. Although Novozym 435 exhibited 30% higher initial activity than MCI-CALB for the butyl acetate synthesis, the reaction course was much more linear using the new preparation, meaning that the MCI-CALB allows for higher productivities per cycle. Both preparations produced around 90% of yield conversions after only 2 h of reaction, using 10% (mass fraction) of enzyme. However, the main advantage of the new biocatalyst was the superior performance during reuse. While Novozym 435 was fully inactivated after only two batches, MCI-CALB could be reused for six consecutive cycles without any washings and keeping around 70% of its initial activity. It is proposed that this effect is due to the higher hydrophobicity of the new support, which does not retain water or acid in the enzyme environment. MCI-CALB has shown to be a very promising biocatalyst for the esterification of small-molecule acids and alcohols.
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, puriss. p.a., ACS reagent