  • Search Results
  • Preparation, characterization, sterility validation, and in vitro cell toxicity studies of microemulsions possessing potential parenteral applications.
Preparation, characterization, sterility validation, and in vitro cell toxicity studies of microemulsions possessing potential parenteral applications.
Drug development and industrial pharmacy (2012-04-07)
Jerry Nesamony, Carrie L Zachar, Rose Jung, Frederick E Williams, Surya Nauli
ABSTRACT
Water-in-oil microemulsions (w/o ME) are ideal for parenteral drug delivery. However, no such formulations have been tested for biocompatibility in in vitro cell cultures. Furthermore, sterilization of w/o MEs is a challenging process that has not been previously developed and validated. To formulate pharmaceutically relevant water-in-oil (w/o) microemulsion's systems suitable for use as a parenteral formulation. w/o MEs were prepared using dioctyl sodium sulfosuccinate (DOSS), ethyl oleate (EO), and water. Formulations were characterized using polarized light microscopy, electrical conductivity, rheology, and dynamic light scattering. An aseptic filtration method for sterilization was developed using membrane filtration. The biocompatibility of selected MEs were evaluated in NIH3T3 cell cultures. Dissolution studies were performed on microemulsions containing methylene blue to evaluate the drug release profile. The maximum amount of water incorporated in the formulations was 14% w/w. DOSS/EO/water microemulsions exhibited Newtonian flow. Particle sizes for these MEs were less than 30 nm in size. Formulations filtered aseptically were free of bacteria when gram-stained and visualized under a microscope. All MEs showed no toxicity to NIH 3T3 cells. The absence of birefringence and low conductivity values indicated that the formulations were w/o microemulsions. The filtration method of sterilization was validated by the absence of microbial growth on blood agar plates over a 14-day period. In vitro dye release studies demonstrate sustained release of the model drug over a 72-h time period. Characteristics delineated in this study demonstrate the potential for these formulations to be used as parenteral preparations.
Sigma-Aldrich
Dioctyl sulfosuccinate sodium salt, ≥97%
Sigma-Aldrich
Docusate sodium, meets USP testing specifications
Sigma-Aldrich
Docusate sodium salt, BioXtra, ≥99%
Sigma-Aldrich
Docusate sodium salt, purum, ≥96.0% (TLC)
Sigma-Aldrich
Ethyl oleate, 98%
Sigma-Aldrich
Docusate sodium salt, BioUltra, ≥99.0% (TLC)
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl oleate, natural, ≥85%
Sigma-Aldrich
Ethyl oleate, technical grade, 70%